ENA428683650 Enamine

1-[1-(4-methyl-1,3-thiazol-2-yl)ethyl]piperazine

  • MDL number MFCD18335012
  • Cas Number : 1267009-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186858

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186858
Availability details for
Quantity
  • Availability
  • Add to Cart