ENA428683653 Enamine

2-(2-bromoethyl)-1,4-dichlorobenzene

  • MDL number MFCD17223175
  • Cas Number : 40173-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186869
Availability details for
Quantity
  • Availability
  • Add to Cart