ENA428683659 Enamine

N-(2-methoxyethyl)-2-methyloxolan-3-amine

  • MDL number MFCD24182100
  • Cas Number : 1550254-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186886
Availability details for
Quantity
  • Availability
  • Add to Cart