ENA428683660 Enamine

(2-methoxyethyl)[(4-methyl-1,3-thiazol-5-yl)methyl]amine

  • MDL number MFCD16793208
  • Cas Number : 1248069-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186887

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186887
Availability details for
Quantity
  • Availability
  • Add to Cart