ENA428683661 Enamine

3-methylbut-2-ene-1-thiol

  • MDL number MFCD00068639
  • Cas Number : 5287-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186888

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186888
Availability details for
Quantity
  • Availability
  • Add to Cart