ENA428683669 Enamine

N-cyclopropyl-5-{2,5-dimethyl-1-[(thiophen-2-yl)methyl]-1H-pyrrol-3-yl}-6H-1,3,4-thiadiazin-2-amine

  • MDL number MFCD08859535
  • Cas Number : 924122-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186930
Availability details for
Quantity
  • Availability
  • Add to Cart