ENA428683672 Enamine

2-(1,3-thiazol-5-yl)pyrimidin-4-amine

  • MDL number MFCD23911096
  • Cas Number : 1511717-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185419
Availability details for
Quantity
  • Availability
  • Add to Cart