ENA428683674 Enamine

1-{[(3-fluoropyridin-2-yl)oxy]methyl}cyclopentan-1-amine

  • MDL number MFCD20349335
  • Cas Number : 1344288-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186959

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186959
Availability details for
Quantity
  • Availability
  • Add to Cart