ENA428683675 Enamine

methyl 4,5-dichloro-1H-indole-2-carboxylate

  • MDL number MFCD08277053
  • Cas Number : 952958-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186961
Availability details for
Quantity
  • Availability
  • Add to Cart