ENA428683676 Enamine

2,4,6-trihydroxy-3-methylbenzaldehyde

  • MDL number MFCD24674224
  • Cas Number : 55743-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186965

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186965
Availability details for
Quantity
  • Availability
  • Add to Cart