ENA428683678 Enamine

4-{4-oxo-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-1-yl}benzonitrile

  • MDL number MFCD11526392
  • Cas Number : 1094828-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186975

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186975
Availability details for
Quantity
  • Availability
  • Add to Cart