ENA428683679 Enamine

1-(4-chlorophenyl)-2-methoxyethan-1-amine

  • MDL number MFCD14691072
  • Cas Number : 391233-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186978
Availability details for
Quantity
  • Availability
  • Add to Cart