ENA428683681 Enamine

1-(4-ethylphenyl)imidazolidin-2-one

  • MDL number MFCD23696652
  • Cas Number : 162748-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186984

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186984
Availability details for
Quantity
  • Availability
  • Add to Cart