ENA428683683 Enamine

1-amino-3-(4-fluorophenyl)propan-2-one hydrochloride

  • MDL number MFCD11848417
  • Cas Number : 93102-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-192
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186992
Availability details for
Quantity
  • Availability
  • Add to Cart