ENA428683690 Enamine

6,6-dimethyl-5,6,7,8-tetrahydroquinazolin-2-amine

  • MDL number MFCD20307253
  • Cas Number : 1344202-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185633

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185633
Availability details for
Quantity
  • Availability
  • Add to Cart