ENA428683691 Enamine

3-(propan-2-yl)-2,3-dihydro-1H-indole

  • MDL number MFCD18809031
  • Cas Number : 67932-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185634

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185634
Availability details for
Quantity
  • Availability
  • Add to Cart