ENA428683696 Enamine

(3S)-3-methyl-1-(oxan-4-yl)piperazine

  • MDL number MFCD11858473
  • Cas Number : 1187931-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185644
Availability details for
Quantity
  • Availability
  • Add to Cart