ENA428683697 Enamine

(1-phenyl-1H-imidazol-2-yl)methanamine dihydrochloride

  • MDL number MFCD08704463
  • Cas Number : 1795396-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187163
Availability details for
Quantity
  • Availability
  • Add to Cart