ENA428683710 Enamine

1,1'-dimethyl-1H,1'H-[3,4'-bipyrazol]-5-amine

  • MDL number MFCD24147148
  • Cas Number : 1414568-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185421
Availability details for
Quantity
  • Availability
  • Add to Cart