ENA428683713 Enamine

7-(chloromethyl)-2,2-dimethyl-2,3-dihydro-1-benzofuran

  • MDL number MFCD08060501
  • Cas Number : 115070-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185648

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185648
Availability details for
Quantity
  • Availability
  • Add to Cart