ENA428683721 Enamine

6-methylthieno[2,3-d]pyrimidine-4-thiol

  • MDL number MFCD11908499
  • Cas Number : 25710-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41223

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41223
Availability details for
Quantity
  • Availability
  • Add to Cart