ENA428683727 Enamine

2,2-dimethylcyclopropane-1-carbonitrile

  • MDL number MFCD01321153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52575

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52575
Availability details for
Quantity
  • Availability
  • Add to Cart