ENA428683742 Enamine

6-bromo-4,8-dichloroquinoline

  • MDL number MFCD12192759
  • Cas Number : 1156602-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67720

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67720
Availability details for
Quantity
  • Availability
  • Add to Cart