ENA428683743 Enamine

4-chloro-6-phenoxyquinoline

  • MDL number MFCD12075239
  • Cas Number : 769952-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67737
Availability details for
Quantity
  • Availability
  • Add to Cart