ENA428683744 Enamine

tert-butyl 4-(bromomethyl)-1H-pyrazole-1-carboxylate

  • MDL number MFCD18837735
  • Cas Number : 530144-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69572

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-69572
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart