ENA428683747 Enamine

2-(dimethoxymethyl)oxolane

  • MDL number MFCD17015703
  • Cas Number : 42187-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185654
Availability details for
Quantity
  • Availability
  • Add to Cart