ENA428683749 Enamine

2-[3-(trifluoromethyl)-1H-pyrazol-4-yl]ethan-1-amine

  • MDL number MFCD09924709
  • Cas Number : 1314904-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75767
Availability details for
Quantity
  • Availability
  • Add to Cart