ENA428683750 Enamine

4-bromo-5-nitroisoquinoline

  • MDL number MFCD00234471
  • Cas Number : 58142-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75823

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75823
Availability details for
Quantity
  • Availability
  • Add to Cart