ENA428683753 Enamine

3-bromo-8-fluoroquinoline

  • MDL number MFCD16658736
  • Cas Number : 855477-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78171

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78171
Availability details for
Quantity
  • Availability
  • Add to Cart