ENA428683755 Enamine

2-methoxypyridine-3-sulfonamide

  • MDL number MFCD25980206
  • Cas Number : 1566061-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78555
Availability details for
Quantity
  • Availability
  • Add to Cart