ENA428683756 Enamine

6-methoxypyridine-3-sulfonamide

  • MDL number MFCD08690716
  • Cas Number : 856955-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78562
Availability details for
Quantity
  • Availability
  • Add to Cart