ENA428683785 Enamine

ethyl 4-phenyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD10696320
  • Cas Number : 127572-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-98244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98244
Availability details for
Quantity
  • Availability
  • Add to Cart