ENA428683787 Enamine

5-chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD03409451
  • Cas Number : 372075-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-98285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98285
Availability details for
Quantity
  • Availability
  • Add to Cart