ENA429358130 Enamine

6-nitro-1,1-dioxo-2,3-dihydro-1lambda6-benzothiophene-3-sulfonyl chloride

  • MDL number MFCD28124988
  • Cas Number : 1803585-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135209
Availability details for
Quantity
  • Availability
  • Add to Cart