ENA429358135 Enamine

2-[5-(propan-2-yl)-1,3,4-oxadiazol-2-yl]morpholine

  • MDL number MFCD28124993
  • Cas Number : 1780747-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-158229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158229
Availability details for
Quantity
  • Availability
  • Add to Cart