ENA429358148 Enamine

9-chloro-1H,2H,3H,4H,5H,6H-azepino[4,3-b]indol-1-one

  • MDL number MFCD28125011
  • Cas Number : 1331936-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182865
Availability details for
Quantity
  • Availability
  • Add to Cart