ENA429358152 Enamine

5-bromo-3-(4-methylbenzoyl)thiophen-2-amine

  • MDL number MFCD28125014
  • Cas Number : 1803609-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184317
Availability details for
Quantity
  • Availability
  • Add to Cart