ENA429358154 Enamine

sodium 2-(1H-1,2,3,4-tetrazol-1-yl)ethane-1-sulfonate

  • MDL number MFCD28125019
  • Cas Number : 1701917-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186697
Availability details for
Quantity
  • Availability
  • Add to Cart