ENA429358255 Enamine

(2S)-2-amino-N,3-dimethylbutanamide

  • MDL number MFCD08059112
  • Cas Number : 87105-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-137395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137395
Availability details for
Quantity
  • Availability
  • Add to Cart