ENA429358326 Enamine

1-methyl-3-(oxolan-3-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD28118214
  • Cas Number : 1596889-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158502
Availability details for
Quantity
  • Availability
  • Add to Cart