ENA429358330 Enamine

N,N-dimethyl-4-(morpholin-2-yl)pyrimidin-2-amine

  • MDL number MFCD18381708
  • Cas Number : 1316218-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158676
Availability details for
Quantity
  • Availability
  • Add to Cart