ENA429358336 Enamine

1-ethyl-2-(1H-imidazol-2-yl)piperazine

  • MDL number MFCD22115332
  • Cas Number : 1508973-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-158812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158812
Availability details for
Quantity
  • Availability
  • Add to Cart