ENA429358346 Enamine

tert-butyl 2-{methyl[(morpholin-2-yl)methyl]amino}acetate

  • MDL number MFCD26998468
  • Cas Number : 1540400-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-159082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159082
Availability details for
Quantity
  • Availability
  • Add to Cart