ENA429358383 Enamine

N-(8-fluoro-3,4-dihydro-2H-1-benzopyran-4-ylidene)hydroxylamine

  • Cas Number : 1018978-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-174778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-174778
Availability details for
Quantity
  • Availability
  • Add to Cart