ENA429358387 Enamine

2-amino-5-(propan-2-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD20645361
  • Cas Number : 288149-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175379
Availability details for
Quantity
  • Availability
  • Add to Cart