ENA429358403 Enamine

5-chloro-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD01927183
  • Cas Number : 13771-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256-257
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176817
Availability details for
Quantity
  • Availability
  • Add to Cart