ENA429358405 Enamine

1-[2-(1,3-dioxolan-2-yl)-1,3-thiazol-5-yl]ethan-1-ol

  • MDL number MFCD28118249
  • Cas Number : 1080060-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177303
Availability details for
Quantity
  • Availability
  • Add to Cart