ENA429358412 Enamine

4-ethylpyridine-2-carbonitrile

  • MDL number MFCD15144729
  • Cas Number : 92486-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178352
Availability details for
Quantity
  • Availability
  • Add to Cart