ENA429358414 Enamine

5-(4-bromophenyl)-3-methylpentanoic acid

  • MDL number MFCD17253978
  • Cas Number : 1343827-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178425
Availability details for
Quantity
  • Availability
  • Add to Cart