ENA429358432 Enamine

3-chloro-4-methyl-5-sulfamoylthiophene-2-carboxylic acid

  • MDL number MFCD28118279
  • Cas Number : 1803603-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179489
Availability details for
Quantity
  • Availability
  • Add to Cart